Anti-BHMT Mouse Monoclonal Antibody [clone: 13B65]

Lieferant: US Biological
029987
USBI029987EA 608 EUR
USBI029987
Anti-BHMT Mouse Monoclonal Antibody [clone: 13B65]
Antikörper
Type: Primary
Antigen: BHMT (betaine--homocysteine S-methyltransferase)
Clonality: Monoclonal
Clone: 13B65
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR