Anti-CDK1 Mouse Monoclonal Antibody [clone: 13B148]

Lieferant: US Biological
030070
USBI030070EA 608 EUR
USBI030070
Anti-CDK1 Mouse Monoclonal Antibody [clone: 13B148]
Antikörper
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Monoclonal
Clone: 13B148
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR