Anti-FABP4 Mouse Monoclonal Antibody [clone: 13B285]

Lieferant: US Biological
030207
USBI030207EA 418 EUR
USBI030207
Anti-FABP4 Mouse Monoclonal Antibody [clone: 13B285]
Antikörper
Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: Monoclonal
Clone: 13B285
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR