Anti-MMP1 Mouse Monoclonal Antibody [clone: 13B525]

Lieferant: US Biological
030447
USBI030447EA 655 EUR
USBI030447
Anti-MMP1 Mouse Monoclonal Antibody [clone: 13B525]
Antikörper
Type: Primary
Antigen: MMP1 (matrix metallopeptidase 1 (interstitial collagenase))
Clonality: Monoclonal
Clone: 13B525
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR