Anti-MYL3 Mouse Monoclonal Antibody [clone: 13B549]
Lieferant: US Biological
030471
USBI030471EA 654 EUR
USBI030471
Anti-MYL3 Mouse Monoclonal Antibody [clone: 13B549]
Antikörper
Type: Primary
Antigen: MYL3 (myosin, light chain 3, alkali; ventricular, skeletal, slow)
Clonality: Monoclonal
Clone: 13B549
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human