Anti-CDKN2A Mouse Monoclonal Antibody [clone: 13B596]

Lieferant: US Biological
030518
USBI030518EA 626 EUR
USBI030518
Anti-CDKN2A Mouse Monoclonal Antibody [clone: 13B596]
Antikörper
Recommended dilution: Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:1000; ELISA: 1:10000

Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: Monoclonal
Clone: 13B596
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR