Anti-p63 alpha Mouse Monoclonal Antibody [clone: 13B598]

Lieferant: US Biological
030520
USBI030520EA 655 EUR
USBI030520
Anti-p63 alpha Mouse Monoclonal Antibody [clone: 13B598]
Antikörper
Type: Primary
Antigen: p63 alpha
Clonality: Monoclonal
Clone: 13B598
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR