Anti-PPY Mouse Monoclonal Antibody [clone: 13B600]

Lieferant: US Biological
030522
USBI030522EA 641 EUR
USBI030522
Anti-PPY Mouse Monoclonal Antibody [clone: 13B600]
Antikörper
Type: Primary
Antigen: PPY (pancreatic polypeptide)
Clonality: Monoclonal
Clone: 13B600
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR