Anti-PARL Mouse Monoclonal Antibody [clone: 13B603]

Lieferant: US Biological
030525
USBI030525EA 573 EUR
USBI030525
Anti-PARL Mouse Monoclonal Antibody [clone: 13B603]
Antikörper
Type: Primary
Antigen: PARL (presenilin associated, rhomboid-like)
Clonality: Monoclonal
Clone: 13B603
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR