Anti-PAX4 Mouse Monoclonal Antibody [clone: 13B605]

Lieferant: US Biological
030527
USBI030527EA 640 EUR
USBI030527
Anti-PAX4 Mouse Monoclonal Antibody [clone: 13B605]
Antikörper
Type: Primary
Antigen: PAX4 (paired box 4)
Clonality: Monoclonal
Clone: 13B605
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR