Anti-PEG10 Mouse Monoclonal Antibody [clone: 13B617]

Lieferant: US Biological
030539
USBI030539EA 654 EUR
USBI030539
Anti-PEG10 Mouse Monoclonal Antibody [clone: 13B617]
Antikörper
Type: Primary
Antigen: PEG10 (paternally expressed 10)
Clonality: Monoclonal
Clone: 13B617
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR