Anti-TEC Mouse Monoclonal Antibody [clone: 13B759]

Lieferant: US Biological
030681
USBI030681EA 573 EUR
USBI030681
Anti-TEC Mouse Monoclonal Antibody [clone: 13B759]
Antikörper
Type: Primary
Antigen: TEC (tec protein tyrosine kinase)
Clonality: Monoclonal
Clone: 13B759
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR