Anti-Blood Group ABO Mouse Monoclonal Antibody [clone: 593CT3.2.2]

Lieferant: US Biological
030808
USBI030808EA 649 EUR
USBI030808
Anti-Blood Group ABO Mouse Monoclonal Antibody [clone: 593CT3.2.2]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:200-6400

Type: Primary
Antigen: Blood Group ABO
Clonality: Monoclonal
Clone: 593CT3.2.2
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR