Anti-CD55 Mouse Monoclonal Antibody [clone: 561CT4.2.3]

Lieferant: US Biological
030856
USBI030856EA 667 EUR
USBI030856
Anti-CD55 Mouse Monoclonal Antibody [clone: 561CT4.2.3]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: CD55 (CD55 molecule, decay accelerating factor for complement (Cromer blood group))
Clonality: Monoclonal
Clone: 561CT4.2.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR