Anti-NCAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 321CT11.4.4]

Lieferant: US Biological
030858-APC
USBI030858-APCEA 884 EUR
USBI030858-APC
Anti-NCAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 321CT11.4.4]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 321CT11.4.4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR