Anti-FUCA2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 234CT20.2.1]
Lieferant: US Biological
030973-APC
USBI030973-APCEA 884 EUR
USBI030973-APC
Anti-FUCA2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 234CT20.2.1]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250
Type: Primary
Antigen: FUCA2 (fucosidase, alpha-L- 2, plasma)
Clonality: Monoclonal
Clone: 234CT20.2.1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human