Anti-FUCA2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 234CT20.2.1]

Lieferant: US Biological
030973-PE
USBI030973-PEEA 884 EUR
USBI030973-PE
Anti-FUCA2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 234CT20.2.1]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250
Do not freeze! Light sensitive

Type: Primary
Antigen: FUCA2 (fucosidase, alpha-L- 2, plasma)
Clonality: Monoclonal
Clone: 234CT20.2.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR