Anti-Fumarase, NY Mouse Monoclonal Antibody [clone: 616CT6.1.6]

Lieferant: US Biological
030974
USBI030974EA 649 EUR
USBI030974
Anti-Fumarase, NY Mouse Monoclonal Antibody [clone: 616CT6.1.6]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1600

Type: Primary
Antigen: Fumarase, NY
Clonality: Monoclonal
Clone: 616CT6.1.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR