Anti-MASTL Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 235CT7.8.2.3]
Lieferant: US Biological
031113-HRP
USBI031113-HRPEA 884 EUR
USBI031113-HRP
Anti-MASTL Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 235CT7.8.2.3]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50
Type: Primary
Antigen: MASTL (microtubule associated serine/threonine kinase-like)
Clonality: Monoclonal
Clone: 235CT7.8.2.3
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human