Anti-PINK1 Mouse Monoclonal Antibody [clone: 38CT20.8.5]
Lieferant: US Biological
031196
USBI031196EA 649 EUR
USBI031196
Anti-PINK1 Mouse Monoclonal Antibody [clone: 38CT20.8.5]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Type: Primary
Antigen: PINK1 (PTEN induced putative kinase 1)
Clonality: Monoclonal
Clone: 38CT20.8.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human