Anti-SERPINH1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 134CT3.1.1]

Lieferant: US Biological
031246-HRP
USBI031246-HRPEA 884 EUR
USBI031246-HRP
Anti-SERPINH1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 134CT3.1.1]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-200; Immunochemistry: 1:50-100

Type: Primary
Antigen: SERPINH1 (serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1))
Clonality: Monoclonal
Clone: 134CT3.1.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR