Anti-TRAF2 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 214CT16.3.4]
Lieferant: US Biological
031306-AP
USBI031306-APEA 884 EUR
USBI031306-AP
Anti-TRAF2 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 214CT16.3.4]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000; Immunofluorescence: 1:10-50
Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: Monoclonal
Clone: 214CT16.3.4
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human