Anti-TRAF2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 214CT16.3.4]

Lieferant: US Biological
031306-FITC
USBI031306-FITCEA 884 EUR
USBI031306-FITC
Anti-TRAF2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 214CT16.3.4]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1000; Immunofluorescence: 1:10-50

Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: Monoclonal
Clone: 214CT16.3.4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR