Anti-ACAD8 Rabbit Polyclonal Antibody

Lieferant: US Biological
031518
USBI031518EA 649 EUR
USBI031518
Anti-ACAD8 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ACAD8 (acyl-CoA dehydrogenase family, member 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR