Anti-AIDA Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
031720-APC
USBI031720-APCEA 884 EUR
USBI031720-APC
Anti-AIDA Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: AIDA (axin interactor, dorsalization associated)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human