Anti-AKIRIN2 Rabbit Polyclonal Antibody

Lieferant: US Biological
031738
USBI031738EA 667 EUR
USBI031738
Anti-AKIRIN2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: AKIRIN2 (akirin 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR