Anti-AKR1C3 Rabbit Polyclonal Antibody

Lieferant: US Biological
031745
USBI031745EA 649 EUR
USBI031745
Anti-AKR1C3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: AKR1C3 (aldo-keto reductase family 1, member C3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR