Anti-Ammonium transporter Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
031842-APC
USBI031842-APCEA 884 EUR
USBI031842-APC
Anti-Ammonium transporter Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Dot Blot 1:500
Do not freeze!

Type: Primary
Antigen: Ammonium transporter
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR