Anti-Ammonium transporter Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
031842-PE
USBI031842-PEEA 884 EUR
USBI031842-PE
Anti-Ammonium transporter Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Dot Blot 1:500
Do not freeze!
Type: Primary
Antigen: Ammonium transporter
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen