Anti-APC Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
031955-APC
USBI031955-APCEA 884 EUR
USBI031955-APC
Anti-APC Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: APC (adenomatous polyposis coli)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse