Anti-APOBEC4 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
031985-AP
USBI031985-APEA 884 EUR
USBI031985-AP
Anti-APOBEC4 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: APOBEC4 (apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 4 (putative))
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human