Anti-ATP13A2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
032264-AP
USBI032264-APEA 884 EUR
USBI032264-AP
Anti-ATP13A2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: ATP13A2 (ATPase type 13A2)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human