Anti-ATP5S Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: US Biological
032289-HRP
USBI032289-HRPEA 884 EUR
USBI032289-HRP
Anti-ATP5S Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: ATP5S (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit s (factor B))
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human