Anti-ATP6V1H Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
032304-APC
USBI032304-APCEA 884 EUR
USBI032304-APC
Anti-ATP6V1H Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: ATP6V1H (ATPase, H+ transporting, lysosomal 50/57kDa, V1 subunit H)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR