Anti-BACH2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
032374-APC
USBI032374-APCEA 616 EUR
USBI032374-APC
Anti-BACH2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Do not freeze!

Type: Primary
Antigen: BACH2 (BTB and CNC homology 1, basic leucine zipper transcription factor 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR