Anti-BACH2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
032375-BIOTIN
USBI032375-BIOTINEA 884 EUR
USBI032375-BIOTIN
Anti-BACH2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: BACH2 (BTB and CNC homology 1, basic leucine zipper transcription factor 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR