Anti-BACH2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: US Biological
032375-HRP
USBI032375-HRPEA 884 EUR
USBI032375-HRP
Anti-BACH2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: BACH2 (BTB and CNC homology 1, basic leucine zipper transcription factor 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human