Anti-BCO2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
032450-BIOTIN
USBI032450-BIOTINEA 884 EUR
USBI032450-BIOTIN
Anti-BCO2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: BCO2 (beta-carotene oxygenase 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR