Anti-BECN1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
032486-BIOTIN
USBI032486-BIOTINEA 884 EUR
USBI032486-BIOTIN
Anti-BECN1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Dot blot 1:500

Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR