Anti-PTRH2 Rabbit Polyclonal Antibody

Lieferant: US Biological
032527
USBI032527EA 667 EUR
USBI032527
Anti-PTRH2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: PTRH2 (peptidyl-tRNA hydrolase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR