Anti-C2 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
032881-BIOTIN
USBI032881-BIOTINEA 884 EUR
USBI032881-BIOTIN
Anti-C2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: C2 (complement component 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human