Anti-C2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
032881-PE
USBI032881-PEEA 884 EUR
USBI032881-PE
Anti-C2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: C2 (complement component 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human