Anti-CAMP Rabbit Polyclonal Antibody

Lieferant: US Biological
033171
USBI033171EA 649 EUR
USBI033171
Anti-CAMP Rabbit Polyclonal Antibody
Antikörper
  • Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CAMP (cathelicidin antimicrobial peptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR