Anti-PTK7 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
033359-APC
USBI033359-APCEA 884 EUR
USBI033359-APC
Anti-PTK7 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-200; Immunochemistry: 1:50-100; Flow Cytometry:1:10-50
Do not freeze!
Type: Primary
Antigen: PTK7 (protein tyrosine kinase 7)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human