Anti-PTK7 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
033359-BIOTIN
USBI033359-BIOTINEA 884 EUR
USBI033359-BIOTIN
Anti-PTK7 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-200; Immunochemistry: 1:50-100; Flow Cytometry:1:10-50

Type: Primary
Antigen: PTK7 (protein tyrosine kinase 7)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR