Anti-CCNB3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
033395-AP
USBI033395-APEA 884 EUR
USBI033395-AP
Anti-CCNB3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Dot blot 1:500
Do not freeze!
Type: Primary
Antigen: CCNB3 (cyclin B3)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse