Anti-CDK1 Rabbit Polyclonal Antibody

Lieferant: US Biological
033569
USBI033569EA 649 EUR
USBI033569
Anti-CDK1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA1:1000; Western Blot: 1:50-100

Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR