Anti-CHN1 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
033856-BIOTIN
USBI033856-BIOTINEA 884 EUR
USBI033856-BIOTIN
Anti-CHN1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: CHN1 (chimerin 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human