Anti-CMPK2 Rabbit Polyclonal Antibody

Lieferant: US Biological
034029
USBI034029EA 649 EUR
USBI034029
Anti-CMPK2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: CMPK2 (cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR