Anti-DACH1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
034456-APC
USBI034456-APCEA 884 EUR
USBI034456-APC
Anti-DACH1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: DACH1 (dachshund family transcription factor 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse